Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C5a Rabbit Polyclonal Antibody

Supplier: BioVision

5895-100
10007-970EA 252.35 USD InStock
10007-970
Anti-C5a Rabbit Polyclonal Antibody
Antibodies
Mouse complement C5a is a 77-amino acid peptide generated during complement activation from the alpha-chain of complement C5. Mouse C5a shares 60% and 82% amino acid sequence identity to human and rat C5a, respectively. C5a binds to a signaling G-protein coupled receptor (GPCR) (C5aR/CD88), inducing neutrophil chemotaxis and endothelial cell activation. It also triggers an oxidative burst in macrophages and neutrophils, and induces release of histamine in basophils and mast cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: C5a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty