Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-hPL Rabbit Polyclonal Antibody

Supplier: BioVision

5869-100
10007-964EA 252.35 USD InStock
10007-964
Anti-hPL Rabbit Polyclonal Antibody
Antibodies
Human Placental Lactogen (HPL), also called human chorionic somatomammotropin, is a polypeptide placental hormone. Its structure and function is similar to that of growth hormone. It modifies the metabolic state of the mother during pregnancy to facilitate the energy supply of the fetus. Recombinant human Placental Lactogen is a 22.3 kDa protein.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: hPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty