Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HNF4A Rabbit Polyclonal Antibody

Supplier: BioVision

3688-100
10004-248EA 252.35 USD InStock
10004-248
Anti-HNF4A Rabbit Polyclonal Antibody
Antibodies
HNF-4 (Hepatocyte nuclear factor 4), a member of the steroid/thyroid nuclear receptor superfamily, is a transcriptional factor which expresses in the liver, intestine, kidney, and pancreatic cells. It contains several functional domains: a ligand-independent activation domain (AF1), a zinc finger DNA binding domain, and a ligand-dependent activation domain (AF2). HNF-4 binds to a specific DNA element as a homodimer and regulates the expression of many genes, involved in glucose, fatty acid, and cholesterol metabolisms.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HNF4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty