Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COX3 Rabbit Polyclonal Antibody

Supplier: BioVision

3687-100
10004-540EA 252.35 USD InStock
10004-540
Anti-COX3 Rabbit Polyclonal Antibody
Antibodies
COX proteins are membrane-associated heme proteins that have cyclooxygenase and peroxidase activities. These enzymes are targets of NSAID (Nonsteroidal anti-inflammatory drμgs) such as aspirin. Prostaglandins (PGs) formed by the enzymatic activity of COX-1 are primarily involved in the regulation of homeostatic functions throμghout the body, whereas PGs formed by COX-2 primarily mediate pain, fever, and inflammation. COX-1 is constitutively expressed, with particularly high expression in gastrointestinal tissues. COX-2 is induced by cytokines and mitogens and is likely to play a role in inflammatory diseases such as rheumatoid arthritis. In rodents and humans, COX-3 encodes proteins with completely different amino acid sequences than COX-1 or COX-2 and without COX activity. It has been sμggested COX-3 as the key to unlocking the mystery of the mechanism of action of acetaminophen.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: COX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty