Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAK3 Rabbit Polyclonal Antibody

Supplier: BioVision

3686-100
10004-536EA 252.35 USD InStock
10004-536
Anti-PAK3 Rabbit Polyclonal Antibody
Antibodies
The p21-activated kinase (PAK) family of serine/threonine kinases is engaged in multiple cellular processes, including cytoskeletal recognization, MAPK signaling, apoptotic signaling, control of phagocytes, NADPH oxidase and growth factor-induced neurite outgrowth. Research indicates that phosphorylation of serine 139 of PAK3 which is located in the kinase inhibitory domain, affects the kinase activity.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PAK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty