Anti-DVL3 Rabbit Polyclonal Antibody

Supplier: BioVision
3684R-100
10004-526EA 286.04 USD
10004-526
Anti-DVL3 Rabbit Polyclonal Antibody
Dvl1, Dvl2, and Dvl3 are mammalian homologs of Dsh (disheveled) proteins that are involved in Wnt signaling pathways. Upon treatment with Wnt proteins, Dvls become hyperphosphorylated and accumulate in the nucleus. Dvls have been reported to mediate endocytosis and may also play a role in certain cancers.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DVL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR