Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MEF2A Rabbit Polyclonal Antibody

Supplier: BioVision

3683-100
10004-524EA 272.95 USD InStock
10004-524
Anti-MEF2A Rabbit Polyclonal Antibody
Antibodies
MEF2A is a member of MEF2 (myocyte enhancer factor 2) family of transcription factors. P38 phosphorylates MEF2A at Thr312 and Thr319 within the transcription activation domain and these phosphorylation events promote MEF2A –MEF2D heterodimer dependent gene expression.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: MEF2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty