Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TAB1 Rabbit Polyclonal Antibody

Supplier: BioVision

3682-100
10004-522EA 272.95 USD InStock
10004-522
Anti-TAB1 Rabbit Polyclonal Antibody
Antibodies
TAB1 (TAK1 binding protein 1) is an adaptor protein that can activate TAK1 in vivo throμgh phosphorylation of TAK1, a mitogen-activated protein kinase kinase kinase. Once activated TAK1 phosphorylates MAPK kinase MKK4 and MKK3/6, which activate p38 MAPK and JNK, respectively.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: TAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty