Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD-14 Mouse Monoclonal Antibody [clone: biG 10]

Supplier: BioVision

3676-100
10004-518EA 345.05 USD InStock
10004-518
Anti-CD-14 Mouse Monoclonal Antibody [clone: biG 10]
Antibodies
The CD14 glycoprotein, gp 55, is present on most monocytic and macrophages like cell types: monocytes, macrophages, Kupffer cells, pleural phagocytic cells and dendritic reticular cells. CD14 is also observed on granulocytes and activated or transformed B-cells. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids. The CD14 molecule has been reported to be a receptor for endotoxin.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CD-14
Clonality: Monoclonal
Clone: biG 10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Horse, Bovine

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty