Anti-FOXO3 Rabbit Polyclonal Antibody

Supplier: BioVision
3673-100
10004-510EA 286.04 USD
10004-510
Anti-FOXO3 Rabbit Polyclonal Antibody
Mammalian forkhead members of the class O (FOXO) transcription factors, including FOXO1, FOXO3a, and FOXO4, are implicated in the regulation of a variety of cellular processes, including the cell cycle, apoptosis, DNA repair, stress resistance, and metabolism. FOXO proteins are negatively regulated by the phosphatidylinositol 3-kinase-Akt signaling pathway, which is activated by growth factors and cytokines. Recent studies indicate that the activities of FOXO proteins are also regulated by oxidative stress, which induces their phosphorylation, translocation to the nucleus, and acetylation-deacetylation. Similar to the tumor suppressor p53, FOXO is activated by stress and induces the expression of genes that contribute to cell-cycle arrest, sμggesting that it also functions as a tumor suppressor.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FOXO3a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR