Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BIN1 Rabbit Polyclonal Antibody

Supplier: BioVision

3672-100
10004-506EA 252.35 USD InStock
10004-506
Anti-BIN1 Rabbit Polyclonal Antibody
Antibodies
BIN-1 is a tumor suppressor protein that was identified in a genetic screen for proteins that interact with the MCY oncoprotein. BIN-1 inhibits the oncogenic activity of MYC, possibly throμgh activating or facilitating apoptosis. BIN is a short-lived protein with a half-life of about 2 hours.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty