Anti-ANTXR1 Rabbit Polyclonal Antibody

Supplier: BioVision
3668-100
10004-494EA 286.04 USD
10004-494
Anti-ANTXR1 Rabbit Polyclonal Antibody
Tumor Endothelial Markers (TEMs, TEM 1-9) are recently identified genes, whose expression is elevated in tumor-associated endothelium. TEMs are barely detectable in normal endothelial cells. TEM8 has been shown to interact with C5 domain of collagen a3(VI), a protein also preferentially expressed in tumor endothelium. It is thoμght that TEM8 may play a role in tumor angiogenesis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ANTXR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR