Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MST1 + MST2 Rabbit Polyclonal Antibody

Supplier: BioVision

3667-100
10004-490EA 252.35 USD InStock
10004-490
Anti-MST1 + MST2 Rabbit Polyclonal Antibody
Antibodies
MST-1 (Mammalian STE20-like kinase 1) is a serine/threonine kinase that has been implicated to play a role in chromatin condensation. Its C-terminal contains nuclear export signals which are released upon caspase mediated cleavage. The N-terminus portion of MST-1 then translocates to the nucleus and will promote chromatin condensation. The full length of MST-1 is localized to the cytoplasm.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MST1 + MST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty