Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FKBP38 Rabbit Polyclonal Antibody

Supplier: BioVision

3666-100
10004-486EA 252.35 USD InStock
10004-486
Anti-FKBP38 Rabbit Polyclonal Antibody
Antibodies
FK506-binding protein 38 (FKBP38), a member of the immunophilin family, has been implicated to play an important role in apoptosis throμgh its involvement in the mechanism that targets Bcl-2 and Bcl-xL to the outer mitochondrial membrane (OMM). Suppression of endogenous FKBP38 by RNAi or transfection of a mutant FKBK38 missing the transmembrane domain necessary for mitochondrial insertion, resulted in the translocation of Bcl-2 and Bcl-xL from the OMM to the cytosol. It has also been sμggested that FKBP38 may play a role in the cross-talk between the Bcl-2 and the Calcineurin/NF-AT apoptosis signaling pathways and in cell size regulation throμgh interactions with tuberous sclerosis genes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FKBP38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty