Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APOB Goat Polyclonal Antibody

Supplier: BioVision

3661-100
10004-468EA 262.65 USD InStock
10004-468
Anti-APOB Goat Polyclonal Antibody
Antibodies
Apolipoprotein B consists of a single polypeptide chain with a molecular weight of 549 kDa and is mostly synthesised in the liver. It is a major apolipoprotein of very low density, intermediate density and low density lipoproteins (LDL) as well as being a major component of lipoprotein (a). Apolipoprotein B is a ligand for the LDL receptor and elevated levels are associated with premature atherosclerosis. Normal plasma apolipoprotein B levels are around 800 mg/L.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Immunoprecipitation: 5-10 µg/ml; Immunohistochemistry: 5-10 µg/ml; ELISA: 0.5-1 µg/ml

Type: Primary
Antigen: APOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty