Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MATH1 Rabbit Polyclonal Antibody

Supplier: BioVision

3658-100
10004-462EA 288.4 USD InStock
10004-462
Anti-MATH1 Rabbit Polyclonal Antibody
Antibodies
Math1 belongs to the basic helix-loop-helix (BHLH) family of transcription factors. It activates E-box dependent transcription along with E47. It is neural-specific and is switched on during differentiation into neuroectoderm.

Recommended Dilutions: Western Blot: 1:200-1000; Immunohistochemistry: 1:100-500

Type: Primary
Antigen: MATH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty