Anti-LIPG Rabbit Polyclonal Antibody

Supplier: BioVision
AB24447-100
10004-458EA 367.24 USD
10004-458
Anti-LIPG Rabbit Polyclonal Antibody
Antibodies
Endothelial lipase (EL) is a member of the triglyceride lipase gene family. It functions primarily as a phospholipase and has low triglyceride lipase activity. It was originally cloned from endothelial cells and found to be expressed in high level in the liver, placenta, lung, ovary, and macrophage. The wide spread distribution of this protein sμggests it plays a general role in lipid metabolism. Human endothelial lipase has an estimated molecular weight of 57 kDa.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 5-20 µg/ml, frozen & paraffin sections, human thyroid

Type: Primary
Antigen: LIPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR