Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EMR2 Rabbit Polyclonal Antibody

Supplier: BioVision

3652-100
10004-454EA 252.35 USD InStock
10004-454
Anti-EMR2 Rabbit Polyclonal Antibody
Antibodies
EGF-like module-containing mucin-like receptor 2 (EMR2) is a member of the class B seven-span transmembrane (TM7) receptor family. EMR2 is expressed predominantly by cells of the immune system such as leukocytes. EMR2 is proteo-lytically cleaved into two subunits: a seven-transmembrane subunit, and an extracellular α subunit.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: EMR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty