Anti-TYH Rabbit Polyclonal Antibody

Supplier: BioVision
3649-100
10004-450EA 402.79 USD
10004-450
Anti-TYH Rabbit Polyclonal Antibody
Tyrosine hydroxylase play an important role in the synthesis of dopamine and other catecholamines. The enzyme’s activity is regulated in the level of transcription and translation, as well as posttranscriptional modifications. Phosphorylation at Ser40 may play a key role in regulation of the catalytic activity of the enzyme.

Recommended Dilutions: Western Blot: 1:200-1000; Iimmunofluorescence: 1:50-100

Type: Primary
Antigen: TYH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR