Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYR61 Rabbit Polyclonal Antibody

Supplier: BioVision

3648-100
10004-446EA 319.3 USD InStock
10004-446
Anti-CYR61 Rabbit Polyclonal Antibody
Antibodies
Cyr61 is a member of angiogenic and vasculogenic regulators designated CCN proteins, which includes CTGF, Cyr61 homolog, Fisp2. Cyr61 functions as an angiogenic inducer, stimulating cell adhesion and migration and promotes DNA synthesis of human vascular endothelial cells. Elevated expression of Cyr61 is observed during vessel growth, wound healing and chondrocyte differentiation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; and Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: CYR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty