Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADH Rabbit Polyclonal Antibody

Supplier: BioVision

3647-100
10004-444EA 272.95 USD InStock
10004-444
Anti-ADH Rabbit Polyclonal Antibody
Antibodies
ADH (alcohol dehydrogenase) family of proteins metabolizes a variety of substances such as ethanol, retinal, other aliphatic alcohol, hydroxysteroids, and lipid peroxidation products. With the coenzyme NAD, ADH catalyzes the reversible conversion of organic alcohol to ketones or aldehydes. ADH plays a major role in ethanol metabolism.

Recommended Dilutions: Western Blot: 1-4 µg/ml; Iimmunofluorescence: 5-20 µg/ml

Type: Primary
Antigen: ADH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty