Anti-6xHis Mouse Monoclonal Antibody [clone: BV-G2]

Supplier: BioVision
3646-100
10004-442EA 344.42 USD
10004-442
Anti-6xHis Mouse Monoclonal Antibody [clone: BV-G2]
It is expected that the His-Tag monoclonal antibody will react with proteins tagged with His from all species.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunocytochemistry: 10-20 µg/ml

Liquid in phosphate buffered saline (PBS, pH 7.4), with 0.05% sodium azide.

Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: BV-G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: All Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR