Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP10 Rabbit Polyclonal Antibody

Supplier: BioVision

3410-200
10004-036EA 262.65 USD InStock
10004-036
Anti-CASP10 Rabbit Polyclonal Antibody
Antibodies
Caspase family of cysteine proteases play a key role in apoptosis. Caspase-10 is expressed in many tissues and cell lines, and has been identified as caspase-10/a (short form) and caspase-10/b (long form), respectively. Similar as other caspases, caspase-10 is also synthesized as an inactive pro-enzyme that is processed in cells undergoing apoptosis. Activated caspase-10 activates caspase-3,-4,-6,-7,-8, and –9, which are then involved in proteolytic cleavage of many cellular proteins. Caspase-10 has been shown involving in the Fas and TNFR1 induced apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CASP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty