Anti-Bcl-6 Antibody

Supplier: BioVision
AB19011-100
10004-028EA 777.09 USD
10004-028
Anti-Bcl-6 Antibody
Antibodies
Bcl-6 encodes a protein of 706 amino acids with a predicted molecular weight of 79 kDa. The Bcl-6 protein has homology with zinc finger transcription factors. However, little is known about its functions.

Recommended Dilutions: Western blot: 0.5-4 µg/ml; Neutralization: 0.5-5 µg/ml; Immunoprecipation: 10-20 µg/ml; and Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: BCL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR