Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCL3 Rabbit Polyclonal Antibody

Supplier: BioVision

3407-100
10004-024EA 288.4 USD InStock
10004-024
Anti-BCL3 Rabbit Polyclonal Antibody
Antibodies
Bcl-3, a proto-oncogene, is believed to be involved in certain human B cell leukemias. It encodes a protein that functions as an IkB-like molecule for native NFkB, but is specific for the p50 subunit of NFkB.

Recommended Dilutions: Western Blot: 2-4 µg/ml

Type: Primary
Antigen: BCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty