Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Etk/BMX Rabbit Polyclonal Antibody

Supplier: BioVision

3398R-100
10003-910EA 252.35 USD InStock
10003-910
Anti-Etk/BMX Rabbit Polyclonal Antibody
Antibodies
Etk is a member of the Bruton’s tyrosine kinase family. Etk is expressed in a variety of hematopoietic, epithelial and endothelial cells. It participates in multiple signal transduction pathways. Phosphorylation of tyrosine 566 by Src kinase is required for activation of Etk in vivo. In endothelial and epithelial cells, Etk is regulated by FAK throμgh phosphorylation at tyrosine 40.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml;

Type: Primary
Antigen: Etk/BMX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty