Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ERK5 Rabbit Polyclonal Antibody

Supplier: BioVision

3397-100
10003-904EA 272.95 USD InStock
10003-904
Anti-ERK5 Rabbit Polyclonal Antibody
Antibodies
Erk-5 shares high homology in the amino-terminal kinase domain with Erk1/2. However, the long carboxy-terminal kinase domain makes Erk5 a unique MAP kinase. Erk5 is activated by a variety of extracellular signals. The direct upstream kinase of Erk5 is defined as MEK5. Studies show that Erk5 pathway is involved in proliferation, differentiation, oncogenesis, and development.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ERK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty