Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HMOX2 Rabbit Polyclonal Antibody

Supplier: BioVision

3392-100
10003-632EA 288.4 USD InStock
10003-632
Anti-HMOX2 Rabbit Polyclonal Antibody
Antibodies
Heme oxygenase (HO) is a microsomal enzyme that oxidatively cleaves heme, a pro-oxidant, into carbon monoxide and biliverdin. Two heme oxygenase isozymes, termed HO-1 and HO-2, have been identified so far. At the amino acid level, HO-1 and HO-2 are 42% homologous. However, each isozyme is evolutionary highly conserved. HO-1 is the inducible form of the enzyme and is therefore also referred to as stress protein Hsp32. Expression of HO-1 can be induced by a variety of stimuli such as heme, metals, hormones, UV radiation and sulfhydryl depleting agents. In contrast, HO-2 is the constituitive form of the enzyme. It is not inducible by exogeneous stimuli and HO-2 is the most prevalent form in most tissues, except the spleen where HO-1 levels are predominant under normal conditions.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 20 µg/ml; Immunohistochemistry: 20 µg/ml, frozen section tested

Type: Primary
Antigen: HMOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, Hamster

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty