Anti-HMOX1 Rabbit Polyclonal Antibody

Supplier: BioVision
3391-100
10003-628EA 326.9 USD
10003-628
Anti-HMOX1 Rabbit Polyclonal Antibody
Heme oxygenase-1 (HO-1) or HSP32 is the inducible isoform of heme oxygenase which catalyzes the NADPH, O2 and cytochrome P450 reductase dependent oxidation of heme to carbon monoxide, iron and biliverdin that is immediately reduced to bilirubin. To date, three heme oxygenase isoforms HO-1, HO-2 and HO-3 have been identified.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: HMOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey, Hamster, Rabbit,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR