Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Hsp90 Rabbit Polyclonal Antibody

Supplier: BioVision

3389-100
10003-620EA 288.4 USD InStock
10003-620
Anti-Hsp90 Rabbit Polyclonal Antibody
Antibodies
Heat shock proteins (HSPs) are ubiquitously expressed in all organisms. A major function of HSP90 and other HSPs is to act as molecular chaperones. HSP90 forms a complex with glucocorticoid receptor (GR), rendering the non ligand-bound receptor transcriptionally inactive. HSP 90 binds the GR as a heterocomplex composed of either HSP56 or cyclophilin-40, forming an aporeceptor complex. HSP90 also exists as a dimer with other proteins such as p60/sti1 and p23, forming an aporeceptor complex with estrogen and androgen receptors.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 10-20 µg/ml; Immunohistochemistry: frozen sections, 10-20 µg/ml

Type: Primary
Antigen: Hsp90
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty