Anti-ELK1 Rabbit Polyclonal Antibody

Supplier: BioVision
3387-100
10003-616EA 309.39 USD
10003-616
Anti-ELK1 Rabbit Polyclonal Antibody
Elk-1 is a transcription factor that binds the serum response element (SRE) and mediates gene activity in response to serum and growth factors. Elk-1 is phosphorylated by MAP kinase pathways and appears to be a direct target of activated MAP kinase. Biochemical studies indicate that Elk-1 is a good substrate for MAP kinase. The kinetics of Elk-1 phosphorylation and activation correlate with MAP kinase activity. Other studies have shown that Elk-1 (Ser383) is also a target of the stress-activated kinase SAPK/JNK.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ELK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR