Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ARHGDIB Rabbit Polyclonal Antibody

Supplier: BioVision

3385-100
10003-614EA 272.95 USD InStock
10003-614
Anti-ARHGDIB Rabbit Polyclonal Antibody
Antibodies
D4-GDI (GDP dissociation inhibitor) is a negative regulator of the ras-related Rho family of GTPases. Since the Rho GTPases promote cytoskeleytal and membrane changes associated with apoptotic cell death, the removal of the D4-GDI block throμgh its cleavage is important for inducing apoptosis. Caspase-3 cleaves the 28kD mature form of D4-GDI to give a 23kDa and 5kDa size fragment. The 23kDa fragment then translocates to the nucleus. The mechanisms involving cleavage of D4-GDI with apoptosis are not presently known. Activation of the Jun N-terminal kinase, a regulator of apoptosis, may be one of the mechanisms.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: ARHGDIB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty