Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CaMKII Rabbit Polyclonal Antibody

Supplier: BioVision

3383-100
10003-610EA 272.95 USD InStock
10003-610
Anti-CaMKII Rabbit Polyclonal Antibody
Antibodies
CaMKII is a member of calcium/calmodulin-activated kinase, functioning in neural synaptic stimulation and T-cell receptor signaling. CaMKII has catalytic and regulatory domains. The binding of Ca++/calmodulin to its regulatory domain releases its autoinhibitory effect and activates the kinase. The activated CaMKII further autophosphorylates itself at Thr286 to render the kinase constitutively active.

Recommended Dilutions: Western blotting at 1-2 µg/ml

Type: Primary
Antigen: CaMKII
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty