Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CAPN2 Rabbit Polyclonal Antibody

Supplier: BioVision

3372-100
10003-604EA 288.4 USD InStock
10003-604
Anti-CAPN2 Rabbit Polyclonal Antibody
Antibodies
Calpains are cytosolic, calcium-dependent, cysteine proteases that are ubiquitously expressed in mammalian and avian tissues. Calpain-1 and 2 consist of a common small subunit and a large variable subunit. Domains in the large subunit include the amino terminal domain I, the proteinase domain II, domain III, and the EF-hand domain IV. Type I calpain requires micromolar amounts of calcium, while type II calpain requires millimolar amounts of calcium.

Recommended Dilutions: Western Blot: 1:200-1000 dilutions

Type: Primary
Antigen: CAPN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty