Anti-CTSF Rabbit Polyclonal Antibody

Supplier: BioVision
3371-100
10003-600EA 309.39 USD
10003-600
Anti-CTSF Rabbit Polyclonal Antibody
Cathepsin F is a cysteine protease that plays a role in invariant chain processing and major histocompatibility complex class II peptide loading by microphages. Cathepsin F has the substrate specificity similar to that of cathepsins L and S.

Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: CTSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR