Anti-CTSG Rabbit Polyclonal Antibody

Supplier: BioVision
3370-200
10003-894EA 332.64 USD
10003-894
Anti-CTSG Rabbit Polyclonal Antibody
The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in cellular metabolism, participating in various events such as peptide biosynthesis and protein degradation. Cathepsin G expression is restricted to bone marrow and early myeloid cells.

Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: CTSG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR