Anti-Cadherin Rabbit Polyclonal Antibody

Supplier: BioVision
3365R-100
10003-878EA 286.04 USD
10003-878
Anti-Cadherin Rabbit Polyclonal Antibody
Cadherins comprise a family of Ca-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of tissue structure and morphogenesis. Cadherins consist of large extracellular domains characterized by a series of five homologous NH2 terminal repeats. The most distal of cadherins is thoμgh to be responsible for binding specificity, transmembrane domains and carboxy terminal domains. The relative short intracellular domains interact with a variety of cytoplasmic proteins, such as β-catenin, to regulate cadherin function.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunoprecipitation: 10-20 µg/ml; and Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: Cadherin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR