Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cox-2 Rabbit Polyclonal Antibody

Supplier: BioVision

3362R-100
10003-868EA 252.35 USD InStock
10003-868
Anti-Cox-2 Rabbit Polyclonal Antibody
Antibodies
COX proteins are membrane-associated heme proteins that have cyclooxygenase and peroxidase activities. These enzymes are targets of NSAID (Nonsteroidal anti-inflammatory drμgs) such as aspirin. Prostaglandins (PGs) formed by the enzymatic activity of COX-1 are primarily involved in the regulation of homeostatic functions throμghout the body, whereas PGs formed by COX-2 primarily mediate pain, fever, and inflammation. COX-1 is constitutively expressed, with particularly high expression in gastrointestinal tissues. COX-2 is induced by cytokines and mitogens and is likely to play a role in inflammatory diseases such as rheumatoid arthritis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Cox-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty