Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CREB Rabbit Polyclonal Antibody

Supplier: BioVision

3360R-100
10003-858EA 252.35 USD InStock
10003-858
Anti-CREB Rabbit Polyclonal Antibody
Antibodies
Transcription factor CREB binds the cAMP response element (CRE) and activates transcription in response to a variety of extracellular signals including neurotransmitters, hormones, membrane depolarization, and growth or neurotrophic factors. Protein kinase A and the calmodulin-dependent protein kinase CaMKII stimulate CREB phosphorylation at Ser133, a key regulatory site controlling transcriptional activity. Phosphorylation at Ser133 is also mediated by p44/42 MAP kinase, p90RSK, p38 MAP kinase and MSK1. CREB appears to play an important role in learning and memory in both flies and mice.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CREB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty