Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MART-1/Melan-A Rabbit Polyclonal Antibody

Supplier: BioVision

5823-100
10007-958EA 252.35 USD InStock
10007-958
Anti-MART-1/Melan-A Rabbit Polyclonal Antibody
Antibodies
MART-1 (also known as Melan-A) is a melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Six other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, Tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MART-1/Melan-A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty