Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Endostatin Rabbit Polyclonal Antibody

Supplier: BioVision

5799-100
10007-956EA 252.35 USD InStock
10007-956
Anti-Endostatin Rabbit Polyclonal Antibody
Antibodies
Endostatin has been identified as a C-terminal fragment of Collagen type 18, a recently identified member of a family of collagen-like proteins referred to as multiplexin family Endostatin specifically inhibits proliferation of endothelial cells althoμgh it does not affect the proliferation of EOMA cells. Endostatin also potently inhibits angiogenesis and tumor growth. Endostatin has an important role in endothelial cell adhesion and cytoskeletal organization. Endostatin can be found in vessel walls (elastic fibers) and basement membranes. Recombinant Endosatin expressed in yeast causes G1 arrest of endothelial cells, and endostatin treatment results in apoptosis of HUVE and HMVE cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Endostatin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty