Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP1 Rabbit Polyclonal Antibody

Supplier: BioVision

5781-100
10007-950EA 252.35 USD InStock
10007-950
Anti-MMP1 Rabbit Polyclonal Antibody
Antibodies
Matrix metalloproteinases (MMPs) are zinc-dependent endopeptidases that degrade substances within the extracellular matrix. The MMP family includes six different groups of enzymes: collagenases, gelatinases, stromelysins, transmembrane MMPs, matrilysins and others. MMPs are secreted as proenzymes that have to be cleaved in order to be activated. Other MMPs, plasmins as well as other factors activate MMPs. MMPs are thoμght to play an important role in tissue remodeling associated with various physiological and pathological processes. MMP1 cleaves collagens of types I, II, III, VII and X.  MMP1 is thoμght to play an important role in rheumatoid and osteo-arthritis pathogenesis as mediators of cartilage and bone destruction.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty