Anti-INS Rabbit Polyclonal Antibody

Supplier: BioVision
AB286162-100UG
10007-944EA 1008.04 USD
10007-944
Anti-INS Rabbit Polyclonal Antibody
Antibodies
Insulin is important in stimulating the uptake of glucose from blood in the skeletal muscle and adipose tissue. It is secrered by pancreatic β cells throμgh a glucose sensing pathway. Insulin also increases cell permeability to monosaccharides, amino acids and fatty acids. Insulin is first formed as a precursor molecule, preproinsulin which is then processed into proinsulin and finally to the mature insulin hormone. A mature form of insulin has an A chain and a B chain that are connnected by two dilsufide bonds. Defects in insulin are the cause of type-II diabetes mellitus.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: INS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR