Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TFF2 Rabbit Polyclonal Antibody

Supplier: BioVision

5745-100
10007-936EA 252.35 USD InStock
10007-936
Anti-TFF2 Rabbit Polyclonal Antibody
Antibodies
The Trefoil Factor peptides (TFF1, TFF2 and TFF3) are expressed in the gastrointestinal tract, and appear to play an important role in intestinal mucosal defense and repair. TFF2 has been shown to inhibit gastrointestinal motility and gastric acid secretion. Recent data sμggests a potential role for TFF2 in acute and chronic asthma. Human TFF2 is a 12.0 kDa polypeptide of 106 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml

Type: Primary
Antigen: TFF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty