Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLGF Rabbit Polyclonal Antibody

Supplier: BioVision

5739-100
10007-932EA 272.95 USD InStock
10007-932
Anti-PLGF Rabbit Polyclonal Antibody
Antibodies
PlGF-1(also called Placenta Growth Factor-1, PLGF, and PGF) is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, colon and mammary carcinomas. It signals throμgh the VEGFR-1/FLT1 receptor and stimulates endothelial cell proliferation and migration. Human PlGF-1 is a 29.7 kDa disulfide-linked homodimeric protein of two 131 amino acid polypeptide chains.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PLGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty