Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IGFBP5 Rabbit Polyclonal Antibody

Supplier: BioVision

5723-100
10007-928EA 252.35 USD InStock
10007-928
Anti-IGFBP5 Rabbit Polyclonal Antibody
Antibodies
Human IGF-BP5 is a cysteine-rich secreted protein produced by vascular smooth muscle cells. It is the major IGF-binding protein present in bone tissue and helps potentiate the action of IGF-I on smooth muscle cells, fibroblasts or osteoblasts. Studies show that IGF-BP5 acts as a growth inhibitor and pro-apoptotic agent in breast cancer cells. IGF-BP5 overexpressing mice show an increase in neonatal mortality, reduced female fertility, whole body growth inhibition, and retarded muscle development.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IGFBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty