Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IGFBP1 Rabbit Polyclonal Antibody

Supplier: BioVision

5717-100
10007-924EA 252.35 USD InStock
10007-924
Anti-IGFBP1 Rabbit Polyclonal Antibody
Antibodies
Human IGF-BP1 is a cysteine-rich secreted protein expressed in liver, decidual, and kidneys and is the most abundant IGF-BP in amniotic fluid. Levels of IGF-BP1 in serum are lowest after food. IGF-BP1 binds both IGF-I and IGF-II with equal affinity. Phosphorylated IGF-BP1 hinders IGF actions, whereas nonphosphorylated IGF-BP1 is stimulatory.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IGFBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty