Anti-Prolactin Rabbit Polyclonal Antibody

Supplier: BioVision
5688-100
10007-912EA 286.04 USD
10007-912
Anti-Prolactin Rabbit Polyclonal Antibody
Prolactin (also known as mammotrophin, luterotropic hormone, and lutetropin) is a neuroendocrine hormone secreted by the pituitary gland. Its primary function is to promote and maintain lactation during pregnancy and suckling. In addition, Prolactin plays an immune-regulatory role by stimulating the activities of ornithine decarboxylase and protein kinase C, which are important for the proliferation, differentiation, and function of lymphocytes. Recombinant murine Prolactin is a 22.5 kDa globular protein containing 198 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Prolactin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR