Anti-LGALS3 Rabbit Polyclonal Antibody

Supplier: BioVision
5684-100
10007-910EA 286.04 USD
10007-910
Anti-LGALS3 Rabbit Polyclonal Antibody
Lectins are carbohydrate binding proteins that interact with glycoprotein and glycolipids on the surface of animal cells. The Galectins are lectins that recognize and interact with beta-galactoside moieties. Galectin-3 regulates a number of biological processes, including embryogenesis, inflammatory responses, cell progression and metastasis. Galectin-3 can function intracellularly in controlling cell cycle and preventing T-cell apoptosis, and also extracellularly, in activating various cells, including monocytes/macrophages, mast cells, neutrophils, and lymphocytes. Human Galectin-3 is a globular 26.0 kDa protein containing 250 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: LGALS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR