Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cox-4 Rabbit Polyclonal Antibody

Supplier: BioVision

3638-100
10004-418EA 272.95 USD InStock
10004-418
Anti-Cox-4 Rabbit Polyclonal Antibody
Antibodies
Cytochrome c oxidase (COX) is a 13-subunit complex spanning the inner mitochondrial membrane and responsible for the terminal reduction of dioxygen to water in the electron transport chain. The three core catalytic units COX-1, -2, -3 are trans-membrane proteins encoded by the mitochondrial genome, whereas the remaining 10 subunits are nuclear encoded and expressed in a tissue-specific manner. The expression of nuclear and mitochondrial subunits of the mitochondrial respiratory chain is thoμght to be highly coordinated. Cox-4 is believed to regulate COX activity according to the extramitochondrial ATP/ADP ratio.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: Cox-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty